ContactUsLinkComponent
Ptec™ Benchtop Ionizer, Transforming Technologies
Ptec™ Benchtop Ionizer, Transforming Technologies
Catalog # 76209-934
undefined
Ptec™ Benchtop Ionizer, Transforming Technologies
Catalog # 76209-934
Supplier Number:  IN5140
Specifications
  • Description:
    PTEC™ benchtop ionizer
  • Environmentally Preferable:
  • Cat. No.:
    76209-934
  • Electrical:
    100/240 V, 50/60 Hz
Specifications
About this item
This Ptec™ 68 KHz AC benchtop air ionizer is a small, efficient static elimination device with variable fan speed for protecting electronic components and general static protection.
  • Remove static charges quickly and efficiently
  • Perfect for single station tabletop, bench, or desk use
  • Adjustment limiting design locks in correct air flow for each application
  • Variable fan speed
Using AC corona technology to make the air more conductive, Ptec™ ionizers produce a balanced stream of positive and negative air ions that never need calibration. The device requires minimal service and maintenance, and was designed to make it operation as simple as possible. The removable grills and the replaceable filter simplify emitter maintenance and tool downtime. Controls and indicators are prominently located on the front top panel. A feedback circuit constantly checks for errors and will alarm if there are issues. This model produces a 68 KHz AC output of approximately 2200 V, and a continuous stream of balanced air ions.