ISCK03 ≥98% (by HPLC)
Supplier: Avantor
Certificates
About this item
Synonyms: N-(4-(1H-Imidazol-1-yl)phenyl)-4-(tert-butyl)benzenesulphonamide, Stem-Cell Factor/c-Kit inhibitor, N-(4-(1H-Imidazol-1-yl)phenyl)-4-(tert-butyl)benzenesulfonamide
CAS Number:945526-43-2
:
Formula:C₁₉H₂₁N₃O₂S
MDL Number:MFCD12031591
Molecular Weight:355.50 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together