Order Entry
United States
ContactUsLinkComponent
SanDisk® Ultra USB 3.0 Flash Drive, CDW Computer Center
SanDisk® Ultra USB 3.0 Flash Drive, CDW Computer Center
Catalog # MSPP-3712072
SanDisk® Ultra USB 3.0 Flash Drive, CDW Computer Center
Catalog # MSPP-3712072
Supplier Number:  3712072
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    SanDisk Ultra® USB 3.0 Flash Drive
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-3712072
  • Supplier no.:
    3712072

Specifications

About this item

The SanDisk Ultra USB 3.0 flash drive combines faster data speeds and generous capacity in a compact, stylish package. Spend less time waiting and transfer files to the drive up to ten times faster than with a standard USB 2.0 drive. With enough storage capacities, the drive can accommodate your bulkiest media files and documents.

  • Transfer speeds up to 10x faster than standard USB 2.0 drives
  • Transfer speeds up to 5x faster than standard USB 2.0 drives
  • Backward compatible with USB 2.0
  • Secure file encryption and password protection with included SanDisk® SecureAccess™

Dimensions: 56.09×21.59×10.41 mm

Storage temperature: 14 to 158 °F

Operating temperature: 32 to 95 °F