ContactUsLinkComponent
Micro Eyelash, Ted Pella
Micro Eyelash, Ted Pella
Catalog # MSPP-113
Supplier:  TED PELLA INC MS
Micro Eyelash, Ted Pella
Catalog # MSPP-113
Supplier:  TED PELLA INC MS
Supplier Number:  113
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    No.1 superfine eyelash with wooden handle
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-113
Specifications
About this item
No. 1 superfine eyelash is attached to a fine wooden handle for delicate manipulation of ultra-thin sections in the knife boat of an ultramicrotome or wherever delicate teasing or manipulation is required.
  • Excellent taper and strength