ContactUsLinkComponent
Salem Sump™ Tube with Anti-Reflux Valve, Cardinal Health
Salem Sump™ Tube with Anti-Reflux Valve, Cardinal Health
Catalog # 60351-204
Supplier:  Cardinal Health
Salem Sump™ Tube with Anti-Reflux Valve, Cardinal Health
Catalog # 60351-204
Supplier:  Cardinal Health
Supplier Number:  8888266148
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Salem Sump™ Tube with Anti-Reflux Valve
  • Length:
    6.0 mm (18 French)
  • Material:
    PVC
  • Environmentally Preferable:
  • Cat. No.:
    60351-204
  • Supplier no.:
    8888266148
Specifications
About this item
These sterile nasogastric tubes are features with gastric reflux prevention valve.
  • Double lumen PVC tube, one for suction drainage and another for sump vent
  • Radiopaque Sentinel Line™ with Sentinel Eye™ tubing
  • Integral irrigation funnel for irrigating through sump vent lumen
Funnel end of vent lumen can be used to cap five in one adapter.