ContactUsLinkComponent
EZClick™ TUNEL-in situ DNA Fragmentation/Apoptosis Assay Kit, BioVision
EZClick™ TUNEL-in situ DNA Fragmentation/Apoptosis Assay Kit, BioVision
  76312-786
 :  Avantor
 :  
undefined
EZClick™ TUNEL-in situ DNA Fragmentation/Apoptosis Assay Kit, BioVision
  76312-786
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    EZClick™ TUNEL – in situ DNA Fragmentation/Apoptosis Assay Kit
  • Environmentally Preferable:
  • Cat. No.:
    76312-786
  • No. of assays/kit:
    100
Non-antibody-based assay for the detection of apoptotic cells.
  • Specific, sensitive, compatible with flow cytometry and fluorescence microscopy instruments
  • Does not require antibodies
  • Avoides lengthy incubation times and does not require washes
Apoptosis is a controlled phenomenon in which apoptotic cells are characterized by changes in nucleus (i.e. chromatin condensation, DNA fragmentation), cytoplasm shrinkage and production of apoptotic bodies. These are subsequently phagocytosed by macrophages, parenchymal cells, or neoplastic cells. DNA strand breaks can be analyzed by TUNEL (Terminal deoxynucleotidyl transferase dUTP nick end-labeling) assays.
BioVision’s EZClick™ TUNEL-DNA fragmentation/apoptosis assay utilizes modified EdUTP nucleotides which are incorporated at the 3’-OH ends of fragmented DNA by terminal deoxynucleotidyl transferase (TdT) enzyme and detected based on a click reaction. This assay enables easy delivery and incorporation of EdUTP coupled with high selectivity of detection of DNA damage. The assay is performed under mild reaction conditions which preserve cell morphology, thus enabling the identification of damaged DNA by FACS or fluorescence microscopy. Our assay is fast and capable of detecting a higher percentage of apoptotic cells than the antibody based methods. The kit contains sufficient reagents to detect total/fragmented DNA in apoptotic cells in a 1×96-well plate or on 50 cover slips.