Order Entry
United States
ContactUsLinkComponent
7701™ Prism® Primer, Loctite, Henkel
7701™ Prism® Primer, Loctite, Henkel
Catalog # 37001-751
Supplier:  Loctite
7701™ Prism® Primer, Loctite, Henkel
Catalog # 37001-751
Supplier:  Loctite
Supplier Number:  135449
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Volume:
    1.75 oz.
  • Color:
    Colorless
  • Description:
    Primer Pris med 7701
  • Environmentally Preferable:
  • Cat. No.:
    37001-751
  • Supplier no.:
    19886

Specifications

About this item

This primer is used to make polyolefin and other low energy surfaces suitable for bonding with cyanoacrylate adhesives.

  • Colorless
  • Solvent-based liquid primer

It is only recommended for difficult-to-bond substrates which include PE, PPL, PTFE and thermoplastic rubber materials. It is not recommended where high peel strength is required. It is suitable for use in the assembly of disposable medical devices.

Certifications: ISO 10993 approved.