To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Competitive
- Host:
- Primary antibody reactivity:
- Target protein:4-Hydroxynonenal (4-HNE)
- Description:4-Hydroxynonenal ELISA Kit
- Environmentally Preferable:
- Cat. No.:76305-634
- No. of tests:100 assays
Specifications
About this item
A competitive ELISA kit for the quantitative measurement of 4-Hydroxynonenal in biological fluids.
- Easy, convenient and time-saving method
- Detection Range: 31.25 to 2000 pg/ml
- Sensitivity: <18.75 pg/ml
- Assay precision: Intra-assay: CV <8%, inter-assay: CV <10%
- No significant cross-reactivity or interference was observed
4-hydroxynonenal (4-HNE) is a lipid peroxidation product derived from oxidized ω-6 polyunsaturated fatty acids such as arachidonic acid. 4-HNE is widely used as a marker of lipid peroxidation.
It is also in higher quantities during oxidative stress due to the increase in the lipid peroxidation chain reaction. It exhibits various biological activities such as cytotoxicity, growth inhibiting activity, genotoxicity, and chemotactic activity.
It also inhibits Sirtuin 3 (SIRT3) via thiol-specific modification.The 4-Hydroxynonenal ELISA kit is a competitive ELISA assay for the quantitative measurement of 4-HNE in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of 4-HNE captured from the samples.