ContactUsLinkComponent
UVP iBox® Studio touch, in vivo Imaging Systems, Analytik Jena US
UVP iBox® Studio touch, in vivo Imaging Systems, Analytik Jena US
Supplier:  Analytik Jena US
Certificates
undefined
UVP iBox® Studio touch, in vivo Imaging Systems, Analytik Jena US
Supplier:  Analytik Jena US
Specifications
  • Catalog No:
  • 76299-404
  • 76299-400
  • Filters:
  • 5-Position Automated Filter Wheel
  • 5-Position Automated Filter Wheel
  • Lighting:
  • RGBW Overhead LEDs
  • RGBW Overhead LEDs
  • Image Area:
  • 168×210 mm
  • 168×210 mm
  • Wired/Wireless:
  • Wireless Network Capable
  • Wireless Network Capable
  • Electrical:
  • 115 V
  • 115 V