Immunomagnetic isolation of untouched human T cells.
- Fast, easy-to-use and column-free
- Up to 98% purity with high recovery
- Untouched, viable cells
The EasySep™ Human T Cell Isolation Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product can be used in place of the EasySep™ Human T Cell Enrichment Kit for even faster cell isolations.