To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Strep-Tactin® Superflow® 50% Suspension
- Size:50 ml
- Environmentally Preferable:
- Cat. No.:MSPP-21206025
- Supplier no.:2-1206-025
Specifications
About this item
Strep-Tactin® Superflow® is used for purification of recombinant Strep-tag® proteins under gravity flow and low pressure or FPLC.
Binding capacity: 1 ml sedimented resin (corresponding to 2 ml of a 50% suspension) is useful for the one-step purification of 50 to 100 nmol recombinant Strep-tag® protein (up to 3 mg in case of a 30 KDa protein).