Pluronic® F-68 Polyol solution 10%
Catalog # IC092750049
Supplier: MP Biomedicals
CAS Number:
Specifications
- Size:100 ml
- Environmentally Preferable:
- Cat. No.:IC092750049
Specifications
About this item
Synonyms: Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), Polyoxyethylene-polyoxypropylene block copolymer, PEG-PPG-PEG
:
:8400 g/mol
Storage Temperature:Ambient
CAS Number:9003-11-6
MDL Number:MFCD00082049