Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
Barbed Tees, Newage Industries
Barbed Tees, Newage Industries
Barbed Tees, Newage Industries
Catalog # 76240-494
Supplier:
NewAge Industries
Small Business Enterprise
Expand image
Barbed Tees, Newage Industries
Catalog # 76240-494
Supplier:
NewAge Industries
Supplier Number:
502-6693
Small Business Enterprise
Specifications
Color:
Natural
Material:
PP
Size:
1
/
4
×
3
/
8
×
3
/
8
"
Environmentally Preferable:
Cat. No.:
76240-494
Specifications
About this item
Product Details
Product Details