ContactUsLinkComponent
IncuCyte® Mouse IgG1 FabFluor pH Red Antibody Labeling Reagent, IncuCyte, Essen Biosciences
IncuCyte® Mouse IgG1 FabFluor pH Red Antibody Labeling Reagent, IncuCyte, Essen Biosciences
Catalog # MSPP-472350
Supplier:  Avantor
CAS Number:  
IncuCyte® Mouse IgG1 FabFluor pH Red Antibody Labeling Reagent, IncuCyte, Essen Biosciences
Catalog # MSPP-472350
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    IncuCyte® Mouse IgG1 FabFluor pH Red Antibody Labeling Reagent
  • Size:
    50 µg
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-472350
  • No. of wells:
    96
Specifications
About this item
The IncuCyte® mouse IgG1 FabFluor pH red antibody labeling reagent utilizes a novel pH-sensitive dye coupled antibody fragment, which labels Fc-containing mouse IgG1 test antibodies and allows for rapid antibody internalization and kinetic high-throughput analysis.
  • One vial (50 µg) of lyophilized conjugate sufficient for labeling large quantities of mouse (igG1) test antibodies in 96-well format
The FabFluor reagent is specifically formulated for use in the IncuCyte ® live-cell analysis system and can be added directly to tissue culture wells using a no wash, simple mix and read protocol to acquire live cell images of adherent and non-adherent cells. When labeled antibodies are added to cells, a fluorogenic signal is quickly observed as the Fab-Ab complex is internalized and processed via acidic lyosomes and endosomes. Antibody Internalization can then be monitored using live cell imaging, and quantified using IncuCyte® software and analysis system.