ContactUsLinkComponent
Connector MIL-DTL-26500, DMC
Connector MIL-DTL-26500, DMC
Catalog # 76251-500
Supplier:  DMC TE
Connector MIL-DTL-26500, DMC
Catalog # 76251-500
Supplier:  DMC TE
Supplier Number:  CM837-16AS
Specifications
  • Description:
    Connector Straight with Normal, 6, 8 keying position, 16 Shell
  • Environmentally Preferable:
  • Cat. No.:
    76251-500
Specifications
About this item
This is with threaded coupling.
  • Aluminium or steel shell
  • Green in color
Insert rotation in positions 1 through 5 minor key rotations, in positions 6 through 10 rotations.