ContactUsLinkComponent
Double Coated Paper Tape 410M, 3M
Double Coated Paper Tape 410M, 3M
Catalog # 76245-926
Supplier:  3M
Double Coated Paper Tape 410M, 3M
Catalog # 76245-926
Supplier:  3M
Supplier Number:  7010373939
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Width:
    1"
  • Description:
    Double Coated Paper Tape
  • Length:
    10 yds.
  • Thickness:
    5 mil.
  • Environmentally Preferable:
  • Cat. No.:
    76245-926
Specifications
About this item
Double Coated Paper Tape 410M is a natural rubber double coated tape with heavy paper carrier for increased strength and dimensional stability.
  • Offers good initial adhesion to a variety of surfaces
  • For indoor and room temperature applications
  • For core starting, end tabbing of papers, films and foils
Excellent balance of high initial adhesion and good holding power to a wide variety of materials. The paper carrier has been specially treated to improve its strength and removability characteristics from high internal strength surfaces.