Order Entry
United States
ContactUsLinkComponent
 
Horizon Horizontal Centrifuge, HORIZON 6 Flex FA, Drucker Diagnostics
  76300-130
 :  Avantor
Irregular Voltage
undefined
Horizon Horizontal Centrifuge, HORIZON 6 Flex FA, Drucker Diagnostics
  76300-130
 :  Avantor
Irregular Voltage

 

  • Description:
    HORIZON 6 Flex FA Fixed Angle Rotor Centrifuge
  • Environmentally Preferable:
  • Cat. No.:
    76300-130
  • W×D×H:
    12×14×9" (30×36×23 cm)
  • Electrical:
    95 - 253 V, 50/60 Hz
  • Max. speed:
    3900 rpm
  • Weight:
    5.4 kg (12 lbs.)

 

 

The HORIZON 6 Flex FA centrifuge features a fully digital display and 10 programmable cycles for flexible fixed angle processing in the routine lab. Easily save and select your custom time, RPM, and g-force settings, or choose from three carefully formulated presets. The LED lid lighting prevents forgotten samples and indicates when the centrifuge is ready, running, or done.

  • Lid lighting indicates cycle status
  • Separate specimens at a fixed angle of 45 degrees
  • Provides maximum safety and durability
  • Allows for safe sample observation and optical calibrations