Order Entry
United States
ContactUsLinkComponent
Miniature Nylon Brush, Solo Horton Brushes
Miniature Nylon Brush, Solo Horton Brushes
Catalog # 76240-934
Miniature Nylon Brush, Solo Horton Brushes
Catalog # 76240-934
Supplier Number:  1104N

Specifications

  • Bristle material:
    Nylon
  • Brush head Ø:
    0.030 mm
  • Handle material:
    Wire
  • Total length:
    3"
  • Environmentally Preferable:
  • Cat. No.:
    76240-934
  • Brush head length:
    1/4"

Specifications

About this item

Miniature tube brushes are ideal for use on precision components.

  • Manually operated
  • With wire handle

Typical users include electronic manufacturers, scientists and product designers.