Braid-Reinforced Silicone Tubing STHT-R, Holland Applied Technologies
Supplier: Avantor
Certificates
About this item
Braid-Reinforced Silicone Tubing with type connectors STHT-R.
- Flexible
- Sterilizable, autoclavable, extremely durable
- Imparts no taste or Odors
- Product code marking
- High pressure rating
The inliner is created exclusively from platinum-catalylized silicone. Continuous extrusion of the inliner allows a smooth internal surface and the most optimum cleaning and sterilisation results. The material reinforcement was developed for increased system pressures.
Frequently Bought Together