To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:SOXA
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Shipping Temperature:Ambient
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
- Cat. No.:75935-232
- Supplier no.:200106093-0100
Specifications
About this item
Sarcosine Oxidase is an inducible enzyme that catalyzes the oxidative demethylation of sarcosine to glycine, hydrogen peroxide, and 5,10-methylene-tetrahydrofolate in the presence of tetrahydrofolate and oxygen. Sarcosine oxidase is also able to oxidize cyclic amino acids such as L-proline and L-pipecolic acid, albeit at reduced rates.
Anti-Sarcosine Oxidase antibody has been assayed against 1.0 ug of Sarcosine Oxidase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.
Type: Primary
Antigen: SOXA
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity: