Order Entry
United States
ContactUsLinkComponent
Anti-cas9 Rabbit Polyclonal Antibody (BAC)
Anti-cas9 Rabbit Polyclonal Antibody (BAC)
  76262-844
undefined
Anti-cas9 Rabbit Polyclonal Antibody (BAC)
  76262-844
 :  600-406-GK0S
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    cas9
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western Blot:
    Yes
  • Size:
    25 µL
  • Environmentally Preferable:
  • Shipping Temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. No.:
    76262-844

 

 

The Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR) and CRISPR Associated (Cas) system was discovered in bacteria as a defense against foreign DNA, even being believed to act as a sort of prokaryotic immune system. The Cas9 protein is becoming a a useful tool in the field of genomic editing for its ability to induce site-directed double strand breaks in DNA. It can cleave almost any sequence complementary to the guide RNA and has recently been used for modifying the genome of human embryos for the first time.

Cas9 Biotin Conjugated Antibody has been tested for use in ELISA, Immunofluorescence, and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band ~158 kDa in size corresponding to Cas-9 protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: cas9
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Rabbit
Isotype:
Reactivity: