Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bags
Sample Bags
Zipper Bags, Cleanroom Film and Bags
Zipper Bags, Cleanroom Film and Bags
76239-470
:
CLEANROOM FILM & BAGS TE
Zipper Bags, Cleanroom Film and Bags
76239-470
:
CLEANROOM FILM & BAGS TE
:
Z100-2-0606
Color:
Clear
Description:
Zipper bag, Class 100
Dimensions:
6×6"
Material:
Polyethylene
Thickness:
2 mil.
Environmentally Preferable:
Cat. No.:
76239-470