This blocking buffer is specifically formulated to achieve superior reproducible western blotting images using fluorescent systems.
- Concentrated solution
- Fluorescence technology is widely used to detect proteins.
- However, many common visible fluorophores often result in considerable background fluorescence in the visible range.
Visible fluorophores are rarely used for membrane-based protein detection because of this high background. IRDye™800, IRDye™700DX, Alexa Fluor® 680 and Cy5.5™ antibody and reagent conjugates are specifically designed for protein detection methods that use longer-wavelength, near-infrared (IR) fluorophores to visualize proteins in western blotting and other applications. IRDye™800 and IRDye™700DX fluoresce outside the range of most autofluorescence and therefore specific signal is sharply evident from any background giving the best possible signal-to-noise ratio. Detection levels in the picogram range on Western blots rival the sensitivity of chemiluminescence on film. IRDye™800 conjugates are also suitable for immunofluorescence microscopy using commercially available excitation/emission filters in the 780nm/820nm range. Dual simultaneous labeling in western blots or microscopy is achieved when IRDye™800 conjugates are used in conjunction with IRDye™700 or Cy5.5™ conjugates. IRDye™800 and IRDye™700DX conjugates provide an ultra-sensitive and convenient alternative to standard chemiluminescent protein detection methods, as well as a valuable tool for multicolor imaging. Once reacted with the membrane and dried, IRDye™800 and IRDye™700DX conjugated antibody-protein complexes are very stable, and membranes can be stored protected from light, re-washed and/or rescanned.
Application Note: Fluorescence technology is widely used to detect proteins in both the visible and near-infrared ranges. This product allows for superior signal detection and lower background noise when fluorochrome conjugated antibodies are used to visualize proteins in western blotting and other applications. Antibody conjugates prepared with IRDye® 800 and IRDye® 700DX (Licor), Cy2™, Cy3™, Cy3.5™, Cy5™ and Cy5.5™ (GE Healthcare), DyLight™405, DyLight™ 549, DyLight™ 649, DyLight™ 680, and DyLight™ 800 (Thermo Fisher/Pierce) and Alexa Fluor® 488, Alexa Fluor® 532, Alexa Fluor® 546, Alexa Fluor® 647 and Alexa Fluor® 680 (Invitrogen/Molecular Probes) have been validated on various platforms using this product with superior results compared to other commercially available products. In the infrared range, where little to no autofluorescence occurs, specific signal is sharply evident from any background giving the best possible signal-to-noise ratio. This allows for detection levels in the picogram range which rivals the sensitivity of chemiluminescence on film for western blotting. Superior results are also seen when this product is used for simultaneous labeling (multiplex) in western blots or microscopy using various fluorochrome combinations for multicolor imaging. Membranes blocked with this product can be dried and are very stable. Membranes that are stored protected from light can be re-washed and/or rescanned.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable: