About this item
Recombinant Human SCF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Stem Cell Factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor known as c-Kit (CD117) and is thought to play a critical role in the maintenance or survival of hematopoietic stem cells. Human SCF shows no activity on murine cells, but murine and rat SCF are active on human cells. Recombinant human SCF is a non-glycosylated protein, containing 165 amino acids, with a molecular weight of 18.4 kDa
Stem Cell Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Stem Cell Factor in immunological assays. Buffer Formulation: 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: