About this item
Recombinant Human LIF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Human Leukemia Inhibitory Factor (LIF) is member of the IL-6 family and is made by a variety of tissues. Human LIF signals through gp130 to activiate STAT3 in human ESCs . In mouse ESCs, LIF promotes self-renewal and pluripotentcy, but in human cells there is no indication that LIF has the same function. Instead, FGF-basic (FGF-2) and Activin A are used to promote self-renewal and pluripotent potential. Recombinant human LIF is a non-glycosylated protein, containing 181 amino acids, with a molecular weight of 19.9 kDa.
Leukemia Inhibitory Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Leukemia Inhibitory Factor in immunological assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: