Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bags
Sample Bags
Bags, Cleanroom Film and Bags
Bags, Cleanroom Film and Bags
Bags, Cleanroom Film and Bags
Catalog # 76238-734
Supplier:
CLEANROOM FILM & BAGS TE
Expand image
Bags, Cleanroom Film and Bags
Catalog # 76238-734
Supplier:
CLEANROOM FILM & BAGS TE
Supplier Number:
F100-2-1624
Specifications
Description:
Bags, class 100
Dimensions:
16×24"
Material:
Polyethylene
Thickness:
2 mil.
Environmentally Preferable:
Cat. No.:
76238-734
Specifications
About this item
Product Details
Product Details