About this item
FADD or Fas-Associated protein with Death Domain is an adaptor molecule that mediates death signaling by the Fas-receptor, tumor necrosis factor receptor and TRAIL-receptor. FADD binds to these receptors via the C-terminus Death Domain which then unmasks the N-terminal effector domain of FADD thereby allowing it to recruit caspase-8 and activate the cysteine protease cascade leading to apoptosis (1). Cells lacking FADD are defective in intracellular double-stranded RNA (dsRNA)-activated gene expression, including production of type I (alpha/beta) interferons and are thus very susceptible to viral infection (2). FADD Protein is ideal for investigators involved in Signaling Proteins, Adaptor Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, and NfkB Pathway research.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: