Order Entry
United States
ContactUsLinkComponent
Novagen® pET-28b(+) DNA, MilliporeSigma
Novagen® pET-28b(+) DNA, MilliporeSigma
Catalog # EM69865-3
Supplier:  MilliporeSigma
CAS Number:  
Novagen® pET-28b(+) DNA, MilliporeSigma
Catalog # EM69865-3
Supplier:  MilliporeSigma
Supplier Number:  69865-3
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    pET-28b(+) DNA
  • Size:
    10 μg
  • Environmentally Preferable:
  • Storage and Handling:
    Standard Handling
  • Storage Temperature:
    ≤-70 °C
  • Cat. No.:
    EM69865-3
  • Supplier no.:
    69865-3

Specifications

About this item

The pET-28a-c(+) vectors carry an N-terminal His Tag®/thrombin/T7 Tag® configuration plus an optional C-terminal His Tag sequence.

  • Sequence is numbered by the pBR322 convention
  • T7 expression region is reversed on the vector map (TB074)

The f1 origin is oriented so that infection with helper phage will produce virions containing single-stranded DNA that corresponds to the coding strand. The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock).