About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CEACAM‑1/CD66a (rR CEACAM‑1/CD66a; Catalog#80224-R08H; Met1-Ser422) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: Clone: 03
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103632-414
- 103630-234
- Antigen Symbol:
- CEACAM1
- CEACAM1
- Antigen Name:
- CEA cell adhesion molecule 1
- CEA cell adhesion molecule 1
- Antigen Synonyms:
- CEACAM‑1|BGPI|BGP1|CD66a|BGP
- CEACAM‑1|BGPI|BGP1|CD66a|BGP
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 03
- 03
- Reactivity:
- Rat
- Rat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient