About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PD-L1 / B7-H1 / CD274 (rM PD-L1 / B7-H1 / CD274; Catalog#50010-M08H; NP_068693.1; Met1-Thr238) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: Clone: 485
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103632-092
- 103629-880
- Antigen Symbol:
- CD274
- CD274
- Antigen Name:
- CD274 molecule
- CD274 molecule
- Antigen Synonyms:
- PDL1|B7-H1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
- PDL1|B7-H1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 485
- 485
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient