About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD62L / L-Selectin / SELL (rh CD62L / L-Selectin / SELL; Catalog#11838-H08H; P14151-1; Met1-Asn332). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103635-784
- Antigen Symbol:
- SELL
- Antigen Name:
- Selectin L
- Antigen Synonyms:
- LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 01
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG1
- ELISA:
- Yes
- Flow Cytometry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient