This antibody was obtained from a rabbit immunized with purified, recombinant Human TGM2 / Transglutaminase 2 (rh TGM2 / Transglutaminase 2; Catalog#11095-H07B; NP_004604.2; Met 1-Ala 687).
Type: Primary
Antigen: TGM2
Clonality: Monoclonal
Clone: 013
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 103619-420
- Antigen Symbol:
- TGM2
- Antigen Name:
- Transglutaminase 2
- Antigen Synonyms:
- TG2|GNAH|G-ALPHA-h|TGC|HEL-S-45
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 013
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient