About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TREM-1 / TREM1 (rh TREM-1 / TREM1; Catalog#10511-H08H; NP_061113.1; Met1-Arg200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: TREM-1
Clonality: Monoclonal
Clone: 04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103617-702
- Antigen Symbol:
- TREM1
- Antigen Name:
- Triggering receptor expressed on myeloid cells 1
- Antigen Synonyms:
- CD354|TREM-1
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 04
- Reactivity:
- Human
- Host:
- Mouse
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient