This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-4R / CD124 (rh IL-4R / CD124; Catalog#10402-H08H; NP_000409.1; Met1-His232) and was produced using recombinant antibody technology.
Type: Primary
Antigen: IL-4R/CD124
Clonality: Monoclonal
Clone: R401
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 103649-120
- 103649-118
- Antigen Symbol:
- IL4R
- IL4R
- Antigen Name:
- Interleukin 4 receptor
- Interleukin 4 receptor
- Antigen Synonyms:
- IL4R|IL-4RA|IL4RA|CD124
- IL4R|IL-4RA|IL4RA|CD124
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- R401
- R401
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Neutralisation:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in Histidine and Arginine buffer containing 120 mM NaCl, 0.02% Tween 80, pH 6.0
- 0.2 μm filtered solution in Histidine and Arginine buffer containing 120 mM NaCl, 0.02% Tween 80, pH 6.0
- Shipping Temperature:
- Ambient
- Ambient