About this item
This antibody was a humanized antibody obtained from a mouse immunized with the recombinant Human PD1 / PDCD1 Protein (Catalog#10377-H08H; NP_005009.2; Met 1-Gln 167) and was produced using recombinant antibody technology.
Type: Primary
Antigen: PD1 / PDCD1 / CD279 Neutralizing
Clonality:
Clone: HN94
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Human IgG4
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103648-826
- 103648-824
- Antigen Symbol:
- PDCD1
- PDCD1
- Antigen Name:
- programmed cell death 1
- programmed cell death 1
- Antigen Synonyms:
- PD-1|hPD-1|hPD-l|CD279|hSLE1|PD1|SLEB2
- PD-1|hPD-1|hPD-l|CD279|hSLE1|PD1|SLEB2
- Antibody Type:
- Primary
- Primary
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- HN94
- HN94
- Reactivity:
- Human
- Human
- Isotype:
- IgG4
- IgG4
- Neutralisation:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in Histidine and Arginine buffer containing 120 mM NaCl, 0.02% Tween 80, pH6.0
- 0.2 μm filtered solution in Histidine and Arginine buffer containing 120 mM NaCl, 0.02% Tween 80, pH6.0
- Shipping Temperature:
- Ambient
- Ambient