About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CHL-1 (rhCHL-1; Catalog#10143-H08H; AAI04919.1; Met 1-Gln 1080). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: CHL1
Clonality: Monoclonal
Clone: 6E6E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103593-610
- Antigen Symbol:
- CHL1
- Antigen Name:
- Cell adhesion molecule L1 like
- Antigen Synonyms:
- LICAM2|L1CAM2|CALL
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 6E6E4
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG1
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient