Syringe Barrel Pistons, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
Certificates
About this item
A piston is inserted into the syringe barrel after it has been loaded with fluid to ensure uniform dispensing, prevent dripping, and eliminate waste by wiping barrel walls clean as fluid is dispensed.
- White SmoothFlow™ wiper pistons are used with most fluids
- Beige SmoothFlow™ pistons are loose-fitting and used with air-entrapped fluids
- Red SmoothFlow™ pistons are tight-fitting and used with mechanical dispensers
- Orange Flat-walled pistons have a looser fit to prevent “bouncing” when dispensing stringy, air-entrapped fluids
- Blue LV Barrier pistons are for cyanoacrylates and very low viscosity fluids
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.