About this item
O-Rings made of Nitrile rubber. It is the general term for acrylonitrile butadiene terpolymer.
- Heat resistance up to 212 °F (100 °C) with shorter life at 250 °F (121 °C)
- Chemical resistance HFA, HFB and HFC fluids
Tolerance to dilute acids, alkali and salt solutions at low temperatures and resistant to aliphatic hydrocarbons (propane, butane, petroleum oil, mineral oil and grease, diesel fuel, fuel oils) vegetable and mineral oils and greases.