ContactUsLinkComponent
ABD Pads, Dukal
ABD Pads, Dukal
Catalog # 76238-266
Supplier:  Dukal
undefined
ABD Pads, Dukal
Catalog # 76238-266
Supplier:  Dukal
Supplier Number:  5940
Specifications
  • Sterility:
    Non sterile
  • Dimensions:
    5×9"
  • Environmentally Preferable:
  • Cat. No.:
    76238-266
  • Packaging:
    25/bag
Specifications
About this item
Highly absorbent multi-layer dressing pads. Soft non-woven outer facing wicks moisture into the cellulose core that quickly absorbs and disperses fluid.
  • Moisture resistant barrier prevents fluid strike-through
  • All edges are sealed to prevent linting and leakage
Not made with natural rubber latex.