ADC® Adtemp 429 No Contact Thermometer
Catalog # 470309-822
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Description:Adtemp 429
- Environmentally Preferable:
- Cat. No.:470309-822
Specifications
About this item
Accurate readings without probe covers.
- One second readings
- 2 temperature modes; Human and Object
- F/C switchable display
- Large illuminated display
The new Adtemp 429 is a non-contact thermometer featuring an infrared sensor that reads temperature from the middle of the forehead. The easy-to-use unit offers an ideal mix of safety and convenience, providing one second results with minimal risk of cross contamination and no costly probe covers.
Ordering information: We apologize, because of increased demand this item will exhibit a delay in delivery.