GSK-650394
Supplier: Avantor
Certificates
About this item
Synonyms: 2-Cyclopentyl-4-(5-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)benzoic acid
CAS Number:890842-28-1
:
Formula:C₂₅H₂₂N₂O₂
MDL Number:MFCD12828779
Molecular Weight:382.45 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together