Miller® Falcon™ Self-Retracting Lifeline, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Nearly indestructible, the Miller® Falcon™ Self-Retracting Lifeline is built with a high impact-resistant nylon housing that withstands the harsh rigors of the toughest applications. Its smooth, round edges increase durability.
- Unique side payout design reduces wear on the entry guide bushing and cable for smooth operation with less drag
- Smooth, quiet operation; Engineered spring retraction tension is designed not to pull or drag on a worker to improve productivity and reduce fatigue
- Built-in, ergonomically-designed molded handle on the 50' (15 m) and 65' (20 m) cable units provide support and leverage when attaching to anchorage connector
- Dual swivels (unit top and snap hook) included on all cable models to minimize lifeline from twisting during use
- New and improved load indicators identify when the unit needs to be removed from service
- Cable models rated to 310 lbs. (140.6 kg) capacity, or 400 lbs. (181.4 kg) when using a SofStop shock absorber (928LS/18INBK)
- Web models rated for up to 400 lbs. (181.4 kg) worker
Load indicators on web models feature a flag on the webbing. Load indicators on cable models are on the snap hook. Units are repairable and require no annual factory recertification, which lowers the cost of ownership and increases productivity (except in Canada where the Canadian Standards Association requires annual factory recertification).
Certifications: Meets all applicable OSHA 1926.502, ANSI A10.32, ANSI Z359.14 Class B, and CSA Z259.2.2 requirements.
Frequently Bought Together