Order Entry
United States
ContactUsLinkComponent
HTP-1500 Heat Therapy Pump for Animals, Kent Scientific
HTP-1500 Heat Therapy Pump for Animals, Kent Scientific
Catalog # MSPP-HTP1500
HTP-1500 Heat Therapy Pump for Animals, Kent Scientific
Catalog # MSPP-HTP1500
Supplier Number:  HTP-1500

Specifications

  • Flow rate:
    15 gph (57 lph)
  • Width:
    10"
  • Capacity:
    51 oz.
  • Description:
    Heat therapy pump with digital display
  • Length:
    7"
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-HTP1500
  • Supplier no.:
    HTP-1500

Specifications

About this item

The HTP-1500 Heat Therapy Pump allows users to safely warm their animals. This is one of the most accurate localized heat therapy system available.

  • Maintain temperature within 1°F for animal safety
  • Mercury-free system decreases potential hazards
  • Uses tap water for added convenience
  • Includes both click-tite and colder connections
  • Accuracy: ± 2 °F

Three temperature safety limits guard against the possibility of overheating and the Hi Limit switches can be tested without taking the unit apart. This provides bright and easy-to-read display which ensures continuous real-time temperature readings.

Temp. set point range: 75 °F to 107 °F (24 °C to 42 °C)