To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Primary antibody reactivity:
- Environmentally Preferable:
- Sensitivity:0.938 ng/ml (1.56 - 100 ng/ml)
- Cat. No.:76288-304
- No. of tests:96 wells
Sensitive ELISA kit for the quantification of Dopamine in human samples.
- High-sensitivity ELISA enables quantification of low amounts of Dopamine
- Rapid quantitative results in <2 hours
- High-throughput immunoassay measures up to 40 samples in duplicate per 96 well kit
- Pre-coated plates reduce errors and save time
It is used for the quantitative determination of human dopamine in serum, plasma, tissue homogenates, and other biological fluids.
The Dopamine ELISA kit is a colorimetric immunoassay kit with results in <2 hours. Absorbance is read at 450 nm. Measure up to 40 samples in duplicate per 96 well kit.