ContactUsLinkComponent
TSPX2 Innovations Microtome Aligner, Universal, Newcomer Supply
  76278-830
undefined
TSPX2 Innovations Microtome Aligner, Universal, Newcomer Supply
  76278-830
 :  6257TO
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Microtome Aligner, Universal
  • Environmentally Preferable:
  • Cat. No.:
    76278-830
Universal aligner that aligns any microtome with ease.
  • The microtome aligner can work on all microtomes
  • You only need one unit per lab
  • A must for biopsies
The universal microtome aligner is a must for every histology laboratory. This tool can quickly and easily align a microtome or multiple microtomes to a standard alignment.
Now when you have to do recuts on an aligned microtome, it's efficient and you don't shave away precious tissue trying to get a proper section recut. When you attempting a biopsy recut, this tool is worth its weight in gold.