Order Entry
United States
ContactUsLinkComponent
Anti-APP Rabbit Monoclonal Antibody [clone: 6E10]
Anti-APP Rabbit Monoclonal Antibody [clone: 6E10]
Catalog # 76284-952
Supplier:  Enzo Life Sciences
Anti-APP Rabbit Monoclonal Antibody [clone: 6E10]
Catalog # 76284-952
Supplier:  Enzo Life Sciences
Supplier Number:  ABS612-0200
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Amyloid beta
  • Antigen Symbol:
    APP
  • Clonality:
    Monoclonal
  • Clone:
    6E10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    200 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen Synonyms:
    A-beta
  • UniProtKB:
    UniProt# P05067
  • Storage Buffer:
    In PBS containing 0.02% Proclin 300.
  • Storage Temperature:
    –20 °C
  • Shipping Temperature:
    Shipped on Blue Ice
  • Immunogen:
    Amyloid-beta peptide.
  • Purification:
    Protein A affinity purified.
  • Cat. No.:
    76284-952

Specifications

About this item

Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: ELISA, IHC, IP, WB, Electron microscopy

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human