Order Entry
United States
ContactUsLinkComponent
Anti-CD52 Mouse Monoclonal Antibody [clone: YTH 34.5-G2b (Campath-1G)]
Anti-CD52 Mouse Monoclonal Antibody [clone: YTH 34.5-G2b (Campath-1G)]
  76284-776
Anti-CD52 Mouse Monoclonal Antibody [clone: YTH 34.5-G2b (Campath-1G)]
  76284-776
 :  ABS454-0200
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD52
  • Antigen Symbol:
    CD52
  • Clonality:
    Monoclonal
  • Clone:
    YTH 34.5-G2b (Campath-1G)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Rhesus monkey,
    Human
  • Western Blot:
    Yes
  • Size:
    200 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen Synonyms:
    Cambridge pathology 1 antigen|CDw52|Epididymal secretory protein E5|Human epididymis-specific protein 5
  • UniProtKB:
    UniProt# P31358
  • Storage Buffer:
    In PBS containing 0.02% Proclin 300.
  • Storage Temperature:
    –20 °C
  • Shipping Temperature:
    Shipped on Blue Ice
  • Immunogen:
    Human peripheral mononuclear cells enriched for T-cells.
  • Purification:
    Protein A affinity purified.
  • Cat. No.:
    76284-776

 

 

Recombinant monclonal antibody (Mouse IgG1κ).

Recommended Applications: ELISA, Flow Cytometry, IHC (FS), IHC (PS), WB, in vivo Assay, Cytotoxicity studies

Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: YTH 34.5-G2b (Campath-1G)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human, Rhesus monkey