- Volume:800 µl
- Description:All-in-one mini spin columns for DNA extraction
- Environmentally Preferable:
- Cat. No.:MSPP-1920250
- Supplier no.:1920-250
EconoSpin® all-in-one silica membrane mini spin column has been widely used as a stand alone single column format.
- Comes with lid
- Binding capacity of 30 to 40 µg allows to be directly plugged into the existing protocol
Its application covers from DNA to RNA, plasmid to viral and genomic DNA, regular PCR purification to next generation sequencing sample preparation.