Order Entry
United States
ContactUsLinkComponent
 
Anti-IgG Mouse Antibody (RPE (R-Phycoerythrin)) [clone: SB108a]
  103264-914
Anti-IgG Mouse Antibody (RPE (R-Phycoerythrin)) [clone: SB108a]
  103264-914
 :  4700-09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Monoclonal
  • Clone:
    SB108a
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Cynomolgus monkey,
    Rhesus monkey
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Storage Buffer:
    Supplied in PBS/NaN3 and a stabilizing agent
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    Rhesus IgG
  • Tested Applications:
    FLISA
  • Cat. No.:
    103264-914
  • Supplier no.:
    4700-09

 

 

PE Anti-Monkey IgG for use in flow cytometry assays.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: SB108a
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rhesus, Cynomolgus