About this item
Increases specificity and yield of primary PCR amplification products.
- Increases yield and specificity of primary PCR amplification products
- Minimizes formation of poorly matched primer-template complexes
- Add to genomic PCR reactions to destabilize many mismatched primer-template complexes
Explore how Perfect Match PCR enhancer increases the specificity and yield of primary PCR amplification products. When added to genomic in vitro amplification reactions, it destabilizes numerous mismatched primer-template complexes that would otherwise result in a heterogeneous population of amplified molecules.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: